EMAIL THIS PAGE TO A FRIEND
Journal of Biomedical Materials Research Part A
5-Florouracil-loaded poly(lactic acid)-poly(caprolactone) hybrid scaffold: potential chemotherapeutic implant.
PMID 24038786