EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Palladium(III)-catalyzed fluorination of arylboronic acid derivatives.
PMID 24040932
Abstract
A practical, palladium-catalyzed synthesis of aryl fluorides from arylboronic acid derivatives is presented. The reaction is operationally simple and amenable to multigram-scale synthesis. Evaluation of the reaction mechanism suggests a single-electron-transfer pathway, involving a Pd(III) intermediate that has been isolated and characterized.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
905879
[Pd(terpy)(MeCN)][BF4]2, ≥95%
C17H14B2F8N4Pd