EMAIL THIS PAGE TO A FRIEND
Annals of internal medicine
Translating science to improve health: hope for patients with kidney disease at what cost?
PMID 24042371
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7455
Tolvaptan, ≥98% (HPLC), powder
C26H25ClN2O3