EMAIL THIS PAGE TO A FRIEND
Stroke
Aggressive medical therapy alone is not adequate in certain patients with severe symptomatic carotid stenosis.
PMID 24046010
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0001
Atorvastatin calcium salt trihydrate, ≥98% (HPLC)
C33H34FN2O5 · 0.5 Ca · 1.5 H2O