EMAIL THIS PAGE TO A FRIEND
ACS chemical neuroscience
Classics in chemical neuroscience: clozapine.
PMID 24047509
Abstract
Clozapine was the first true breakthrough in schizophrenia treatment since the discovery of chlorpromazine in 1950, effectively treating positive, negative, and some cognitive symptoms, as well as possessing unprecedented efficacy in treatment-resistant patients. Despite over 30 years of intense study, the precise molecular underpinnings that account for clozapine's unique efficacy remain elusive. In this Viewpoint, we will showcase the history and importance of clozapine to neuroscience in general, as well as for the treatment of schizophrenia, and review the synthesis, pharmacology, drug metabolism, and adverse events of clozapine.