EMAIL THIS PAGE TO A FRIEND
Gene
Null genotypes of glutathione S-transferase M1 (GSTM1) and T1 (GSTT1) polymorphisms increased susceptibility to type 2 diabetes mellitus, a meta-analysis.
PMID 24050897