EMAIL THIS PAGE TO A FRIEND
Diagnostic microbiology and infectious disease
HindII and SduI digests of heat-shock protein 70 PCR for Leishmania typing.
PMID 24050933
Abstract
Restriction fragment length polymorphisms of the heat-shock protein 70 gene have been used for discriminating Leishmania species. Here, we validated HindII as a much cheaper alternative to EcoRII and SduI for discriminating Leishmania (Viannia) braziliensis from Leishmania (Viannia) naiffi and an atypical Leishmania (V.) braziliensis group, which was previously not possible.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R3884 EclX I from Enterobacter cloacae 590, Restriction Enzyme