EMAIL THIS PAGE TO A FRIEND
FEBS letters
Polysulfide exerts a protective effect against cytotoxicity caused by t-buthylhydroperoxide through Nrf2 signaling in neuroblastoma cells.
PMID 24055470
Abstract
Polysulfide is a bound sulfur species derived from endogenous H2S. When mouse neuroblastoma, Neuro2A cells were exposed to tert-butyl hydroperoxide after treatment with polysulfide, a significant decline in cell toxicity was observed. Rapid uptake of polysulfides induced translocation of Nrf2 into the nucleus, resulting in acceleration of GSH synthesis and HO-1 expression. We demonstrated that polysulfide reversibly modified Keap1 to form oxidized dimers and induced the translocation of Nrf2. Moreover, polysulfide treatment accelerated Akt phosphorylation, which is a known pathway of Nrf2 phosphorylation. Thus, polysulfide may mediate the activation of Nrf2 signaling, thereby exerting protective effects against oxidative damage in Neuro2A cells.