EMAIL THIS PAGE TO A FRIEND
Journal of dual diagnosis
Does Posttraumatic Stress Disorder (PTSD) Affect Post-Treatment Methamphetamine Use?
PMID 24065875
Abstract
Although trauma is a well-established risk factor for substance use disorders, little is known about the association between posttraumatic stress disorder (PTSD) and treatment outcomes among methamphetamine users. In the present study, we examine the relationship between PTSD and post-treatment methamphetamine use outcomes, hospitalizations, and overall psychiatric impairment. Using data from 526 adults in the largest psychosocial clinical trial of methamphetamine users conducted to date, this study examined: (1) treatment outcomes of methamphetamine users with concomitant PTSD three years after psychosocial treatment for methamphetamine dependence; and (2) PTSD symptom clusters as risk factors for post-treatment relapse to methamphetamine use. PTSD was associated with poorer methamphetamine use outcomes; methamphetamine use frequency throughout the 3-year follow-up was significantly greater among individuals with a PTSD diagnosis, and those with PTSD had more than five times the odds of reporting methamphetamine use in the 30 days prior to the follow-up interview, Addressing these high risk PTSD symptoms and syndromes in methamphetamine users may be helpful as a means of improving treatment outcomes in this population.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V4640
VU0357017 monohydrochloride, ≥98% (HPLC)
C18H27N3O3 · HCl