EMAIL THIS PAGE TO A FRIEND
The American journal of clinical nutrition
Cobalamin supplements for infants: a shot in the cradle?
PMID 24067668
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1428000 Hydroxocobalamin, European Pharmacopoeia (EP) Reference Standard
C62H89CoN13O15P