EMAIL THIS PAGE TO A FRIEND
Anais da Academia Brasileira de Ciencias
Multivariate optimization of an ultrasound-assisted extraction procedure for Cu, Mn, Ni and Zn determination in ration to chickens.
PMID 24068081
Abstract
In this work, multivariate optimization techniques were used to develop a method based on the ultrasound-assisted extraction for copper, manganese, nickel and zinc determination from rations for chicken nutrition using flame atomic absorption spectrometry. The proportions of extracting components (2.0 mol.L-1 nitric, hydrochloric and acetic acid solutions) were optimized using centroid-simplex mixture design. The optimum proportions of this mixture taken as percentage of each component were respectively 20%, 37% and 43%. Variables of method (sample mass, sonication time and final acid concentration) were optimized using Doehlert design. The optimum values found for these variables were respectively 0.24 g, 18s and 3.6 mol.L-1. The developed method allows copper, manganese, nickel and zinc determination with quantification limits of 2.82; 4.52; 10.7; e 9.69 µg.g-1, and precision expressed as relative standard deviation (%RSD, 25 µg.g-1, N = 5) of 5.30; 2.13; 0.88; and 0.83%, respectively. This method was applied in the analytes determination from chicken rations collected from specialized commerce in Jequié city (Bahia State/Brazil). Application of paired t-test at the obtained results, in a confidence level of 95%, does not show significant difference between the proposed method and the microwave-assisted digestion.