EMAIL THIS PAGE TO A FRIEND
Chinese medical sciences journal = Chung-kuo i hsueh k'o hsueh tsa chih
Neurotoxicity and biomarkers of lead exposure: a review.
PMID 24074621
Abstract
Appropriate selection and measurement of lead biomarkers of exposure are critically important for health care management purposes, public health decision making, and primary prevention synthesis. Lead is one of the neurotoxicants that seems to be involved in the etiology of psychologies. Biomarkers are generally classified into three groups: biomarkers of exposure, effect, and susceptibility.The main body compartments that store lead are the blood, soft tissues, and bone; the half-life of lead in these tissues is measured in weeks for blood, months for soft tissues, and years for bone. Within the brain, lead-induced damage in the prefrontal cerebral cortex, hippocampus, and cerebellum can lead to a variety of neurological disorders, such as brain damage, mental retardation, behavioral problems, nerve damage, and possibly Alzheimer's disease, Parkinsons disease, and schizophrenia. This paper presents an overview of biomarkers of lead exposure and discusses the neurotoxic effects of lead with regard to children and adults.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
391352
Lead, powder, −100 mesh, 99.95% trace metals basis
Pb
209708
Lead, powder, −325 mesh, ≥99% trace metals basis
Pb
265888
Lead, wire, diam. 1.0 mm, 99.99% trace metals basis
Pb
695912
Lead, shot, 1-3 mm, 99.995% trace metals basis
Pb
396117
Lead, shot, <2 mm, 99.9% trace metals basis
Pb
GF96512597
Lead, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF87346806
Lead, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF29785898
Lead, foil, 5m coil, thickness 0.1mm, as rolled, 99.95%
Pb
GF60047760
Lead, foil, 250x250mm, thickness 3.0mm, as rolled, 99.95%
Pb
GF74259793
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF24521329
Lead, foil, 100x100mm, thickness 3.0mm, as rolled, 99.99+%
Pb
GF21778397
Lead, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF75368470
Lead, foil, light tested, 100x100mm, thickness 0.025mm, 99.99+%
Pb
GF21166321
Lead, foil, 100x100mm, thickness 3.0mm, as rolled, 99.95%
Pb
GF47404672
Lead, foil, 500x500mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF42741637
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Pb
GF06345479
Lead, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF02017167
Lead, wire reel, 2m, diameter 0.5mm, as drawn, 99.999%
Pb
GF05521160
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF68213538
Lead, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF11259121
Lead, foil, 2m coil, thickness 1.0mm, as rolled, 99.95%
Pb
GF36533505
Lead, foil, 300x300mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF23236477
Lead, foil, 10m coil, thickness 0.1mm, as rolled, 99.95%
Pb
GF43321610
Lead, foil, 2m coil, thickness 1.0mm, as rolled, 99.5%
Pb
GF79027847
Lead, rod, 200mm, diameter 12.7mm, 99.95%
Pb
GF85616130
Lead, wire reel, 1m, diameter 0.5mm, as drawn, 99.999%
Pb
GF66906197
Lead, rod, 1000mm, diameter 6.35mm, 99.95%
Pb
GF22298777
Lead, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF08081670
Lead, wire reel, 2m, diameter 1.0mm, as drawn, 99.99+%
Pb
GF95548281
Lead, foil, 10mm disks, thickness 0.02mm, 99.99+%
Pb