EMAIL THIS PAGE TO A FRIEND
International journal of clinical pharmacology and therapeutics
Correlation of skin toxicity and hypertension with clinical benefit in advanced hepatocellular carcinoma patients treated with sorafenib.
PMID 24075093
Abstract
The purpose of this study was to identify the correlation of skin toxicity and hypertension with clinical benefit in advanced hepatocellular carcinoma (HCC) patients treated with sorafenib by analyzing medical records retrospectively. Data from medical records was statistically analyzed to identify a correlation of skin toxicity and hypertension with treatment response and prognosis in advanced HCC patients who had received sorafenib at the Asan Medical Center from July 2010 to June 2012. This study investigated prognostic factors for overall survival and the correlation between the development of skin toxicities and hypertension. A total of 99 patients receiving sorafenib were included in this study. 29 patients who developed skin toxicities Grade 2 or higher showed significantly longer survival than the 70 patients who developed skin toxicities less than Grade 2 or those without skin toxicity (p = 0.024). However, development of hypertension was not related to survival (p = 0.262). In a multivariate analysis, skin toxicities were found to be good prognostic factors for overall survival (hazard ratio, 0.320; 95% CI, 0.119 - 0.861; p = 0.024) as well as low α-fetoprotein level (hazard ratio, 0.195; 95% CI, 0.076 - 0.500; p = 0.001). On the other hand, no correlation was found between the development of skin toxicities and hypertension (p = 0.109). Skin toxicities that are common adverse reactions in advanced HCC patients treated with sorafenib may be used as surrogate markers for clinical benefit. Therefore, early detection and proper management of these toxicities is crucial for continuing treatment with sorafenib.