EMAIL THIS PAGE TO A FRIEND
Nature
Microbial production of short-chain alkanes.
PMID 24077097
Abstract
Increasing concerns about limited fossil fuels and global environmental problems have focused attention on the need to develop sustainable biofuels from renewable resources. Although microbial production of diesel has been reported, production of another much in demand transport fuel, petrol (gasoline), has not yet been demonstrated. Here we report the development of platform Escherichia coli strains that are capable of producing short-chain alkanes (SCAs; petrol), free fatty acids (FFAs), fatty esters and fatty alcohols through the fatty acyl (acyl carrier protein (ACP)) to fatty acid to fatty acyl-CoA pathway. First, the β-oxidation pathway was blocked by deleting the fadE gene to prevent the degradation of fatty acyl-CoAs generated in vivo. To increase the formation of short-chain fatty acids suitable for subsequent conversion to SCAs in vivo, the activity of 3-oxoacyl-ACP synthase (FabH), which is inhibited by unsaturated fatty acyl-ACPs, was enhanced to promote the initiation of fatty acid biosynthesis by deleting the fadR gene; deletion of the fadR gene prevents upregulation of the fabA and fabB genes responsible for unsaturated fatty acids biosynthesis. A modified thioesterase was used to convert short-chain fatty acyl-ACPs to the corresponding FFAs, which were then converted to SCAs by the sequential reactions of E. coli fatty acyl-CoA synthetase, Clostridium acetobutylicum fatty acyl-CoA reductase and Arabidopsis thaliana fatty aldehyde decarbonylase. The final engineered strain produced up to 580.8 mg l(-1) of SCAs consisting of nonane (327.8 mg l(-1)), dodecane (136.5 mg l(-1)), tridecane (64.8 mg l(-1)), 2-methyl-dodecane (42.8 mg l(-1)) and tetradecane (8.9 mg l(-1)), together with small amounts of other hydrocarbons. Furthermore, this platform strain could produce short-chain FFAs using a fadD-deleted strain, and short-chain fatty esters by introducing the Acinetobacter sp. ADP1 wax ester synthase (atfA) and the E. coli mutant alcohol dehydrogenase (adhE(mut)).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55689 Alcohol Dehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg
A7011
Alcohol Dehydrogenase from Saccharomyces cerevisiae, lyophilized powder (contains buffer salts), ≥300 units/mg protein
A3263
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein
A8656 Alcohol Dehydrogenase from Saccharomyces cerevisiae
457116
Decane, anhydrous, ≥99%
C10H22
30540
Decane, analytical standard
C10H22
30550
Decane, purum, ≥98.0% (GC)
C10H22
442669
Decane, analytical standard
C10H22
D901
Decane, ReagentPlus®, ≥99%
C10H22
30570
Decane, ≥95%
C10H22
359343
Hydroquinidine, 95%
C20H26N2O2
74250
Nonane, analytical standard
C9H20
296821
Nonane, anhydrous, ≥99%
C9H20
N29406
Nonane, ReagentPlus®, 99%
C9H20
442694
Nonane, analytical standard
C9H20
S-003
R(-)-Selegiline solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C13H17N
87718
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
C4H12ClN
74202
Tetramethylammonium chloride, for ion pair chromatography, ≥99.0% (AT)
C4H12ClN
T19526
Tetramethylammonium chloride, reagent grade, ≥98%
C4H12ClN
T3411
Tetramethylammonium chloride solution, for molecular biology
T7505
Tetramethylammonium hydroxide pentahydrate, ≥97%
C4H13NO · 5H2O
87741
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
C4H13NO · 5H2O
235946
Tetramethylammonium iodide, 99%
C4H12IN
02799
Tetramethylammonium sulfate, for ion pair chromatography, ≥99.0% (T)
C8H24N2O4S
87743
Tetramethylammonium sulfate hydrate
C8H24N2O4S · xH2O