EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Occurrence of phthalic acid esters in source waters: a nationwide survey in China during the period of 2009-2012.
PMID 24077254
Abstract
The first nationwide survey of six phthalic acid esters (PAEs) (diethyl phthalate (DEP); dimethyl phthalate (DMP); di-n-butyl phthalate (DBP); butyl benzyl phthalate (BBP); bis(2-ethylhexyl) phthalate (DEHP); di-n-octyl phthalate (DnOP)) in source waters was conducted in China. The results showed these PAEs were ubiquitous in source waters. DBP and DEHP were the most frequently detected with high concentrations ranging nd-1.52 μg/L and nd-6.35 μg/L, respectively. These PAEs concentrations (except DBP) in surface water (rivers, lakes and reservoirs) were generally higher than those in groundwater; DBP had high concentrations in groundwater in Northeast China (Liao River Basin) and North China (Hai River Basin). Their concentrations in the northern regions were generally higher than those in the southern and eastern regions; particularly, in North China. Three short-chain PAEs (DMP, DEP and DBP) were detected with high concentrations in Hai River Basin, Pearl River Basin and Yellow River Basin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42438
Benzyl butyl phthalate, certified reference material, TraceCERT®
C19H20O4
36927
Benzyl butyl phthalate, analytical standard
C19H20O4
308501
Benzyl butyl phthalate, 98%
C19H20O4
442503
Benzyl butyl phthalate, analytical standard
C19H20O4
89353
Density Standard 1191 kg/m3, H&D Fitzgerald Ltd. Quality
C10H10O4
40066 Di-n-butyl phthalate solution, certified reference material, 5000 μg/mL in methanol
88173
Di-n-octyl phthalate, certified reference material, TraceCERT®
C24H38O4
31301
Di-n-octyl phthalate, analytical standard
C24H38O4
80153
Di-n-octyl phthalate, ≥98.0% (GC)
C24H38O4
48560-U
Di-n-octyl phthalate, analytical standard
C24H38O4
53008
Diethyl phthalate, PESTANAL®, analytical standard
C12H14O4
524972
Diethyl phthalate, 99.5%
C12H14O4
W512206
Diethyl phthalate, ≥99%
C12H14O4
41320
Dimethyl phthalate, PESTANAL®, analytical standard
C10H10O4
W508500
Dimethyl phthalate, ≥99%
C10H10O4
525081
Dimethyl phthalate, ≥99%
C10H10O4
07-4770
Dimethyl phthalate, SAJ special grade, ≥99.0%
C10H10O4
40069 Dimethyl phthalate solution, certified reference material, 5000 μg/mL in methanol
80010
Phthalic acid, puriss. p.a., ≥99.5% (T)
C8H6O4
402915
Phthalic acid, ACS reagent, ≥99.5%
C8H6O4
P39303
Phthalic acid, reagent grade, 98%
C8H6O4
376485
Phthalic acid dipotassium salt, 98%
C8H4K2O4
P1088
Potassium hydrogen phthalate, BioXtra, ≥99.95%
C8H5KO4
60359
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
C8H5KO4
14659
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®
C8H5KO4
60360
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
C8H5KO4
33325
Potassium phthalate monobasic, puriss. p.a., buffer substance, ≥99.5%
C8H5KO4
60357
Potassium phthalate monobasic, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
C8H5KO4
179922
Potassium phthalate monobasic, ACS reagent, acidimetric standard
C8H5KO4