EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Phase I study of vismodegib in children with recurrent or refractory medulloblastoma: a pediatric brain tumor consortium study.
PMID 24077351