EMAIL THIS PAGE TO A FRIEND
Zhonghua yi xue yi chuan xue za zhi = Zhonghua yixue yichuanxue zazhi = Chinese journal of medical genetics
[Diagnosis and treatment of isolated methylmalonic acidemia].
PMID 24078577
Abstract
To explore the clinical feature, therapeutic effect and prognosis of isolated methylmalonic acidemia. The clinical characteristics, laboratory findings, treatment and outcome of 40 patients were retrospectively analyzed. The main treatment was a low-protein diet supplemented with L-carnitine and special milk free of leucine, valine, threonine and methionine. Vitamin B12 was also given to cobalamin responders. The patients were followed up every 1-3 months. Mutations in the MUT gene were identified in 30 of 33 patients who had accepted DNA testing. Thirty cases were treated and followed up regularly for from 1 month to 8 years. Eight cases had died, 8 had developed normal intelligence, among whom 4 from newborn screening were asymptomatic. Psychomotor developmental delay and mental retardation were present in 14 cases. The propionylcarnitine level, ratio of propionylcarnitine/acetylcarnitine in blood, methylmalonic acid and methylcitric acid levels in urine have decreased significantly, with the median values reduced respectively from 24.15 (7.92-81.02) μmol/L, 1.08 (0.38-6.01), 705.34 (113.79-3078.60) and 7.71 (0.52-128.21) to 10.50 (3.00-30.92) μmol/L, 0.63 (0.25-2.89), 166.23 (22.40-3322.21) and 3.96 (0.94-119.13) (P < 0.05). The prognosis of isolated methylmalonic acidemia may be predicted with the enzymatic subgroup, age at onset and cobalamin responsiveness. Outcome is unfavorable in neonatal patients and those who were non-responsive to cobalamin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L0399900
Levocarnitine, European Pharmacopoeia (EP) Reference Standard
C7H15NO3