EMAIL THIS PAGE TO A FRIEND
Journal of materials chemistry. C
Fluorescent materials for pH sensing and imaging based on novel 1,4-diketopyrrolo-[3,4-
PMID 24078864
Abstract
New optical pH-sensors relying on 1,4-diketopyrrolo-[3,4-
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A48407
4-Amino-2,6-dichlorophenol, 98%
C6H5Cl2NO