EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Rheumatic heart disease back in the limelight.
PMID 24083308
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31735
Benzathine penicilline G tetrahydrate, VETRANAL, analytical standard
C48H56N6O8S2 · 4H2O