EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Identification of transmembrane domain 3, 4 & 5 residues that contribute to the formation of the ligand-binding pocket of the urotensin-II receptor.
PMID 24084430
Abstract
Urotensin-II (UII), a cyclic undecapeptide, selectively binds the urotensin-II receptor (UT receptor), a G protein-coupled receptor (GPCR) involved in cardiovascular effects and associated with numerous pathophysiological conditions including hypertension, atherosclerosis, heart failure, pulmonary hypertension and others. In order to identify specific residues in transmembrane domains (TM) three (TM3), four (TM4) and five (TM5) that are involved in the formation of the UT receptor binding pocket, we used the substituted-cysteine accessibility method (SCAM). Each residue in the F118((3.20)) to S146((3.48)) fragment of TM3, the L168((4.44)) to G194((4.70)) fragment of TM4 and the W203((5.30)) to V232((5.59)) fragment of TM5, was mutated, individually, to a cysteine. The resulting mutants were then expressed in COS-7 cells and subsequently treated with the positively charged sulfhydryl-specific alkylating agent methanethiosulfonate-ethylammonium (MTSEA). MTSEA treatment resulted in a significant reduction in the binding of (125)I-UII to TM3 mutants L126C((3.28)), F127C((3.29)), F131C((3.33)) and M134C((3.36)) and TM4 mutants M184C((4.60)) and I188C((4.64)). No loss of binding was detected following treatment by MTSEA for all TM5 mutants tested. In absence of a crystal structure of UT receptor, these results identify key determinants in TM3, TM4 and TM5 that participate in the formation of the UT receptor binding pocket and has led us to propose a homology model of the UT receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72945
Ethyl methanesulfonate, certified reference material, TraceCERT®
C3H8O3S