EMAIL THIS PAGE TO A FRIEND
The Annals of thoracic surgery
Invited commentary.
PMID 24088449
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7106
Azelnidipine, ≥98% (HPLC), powder
C33H34N4O6