EMAIL THIS PAGE TO A FRIEND
Drug delivery
Augmented cytotoxicity of hydroxycamptothecin-loaded nanoparticles in lung and colon cancer cells by chemosensitizing pharmaceutical excipients.
PMID 24093513