EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and biological evaluation of 5-carbamoyl-2-phenylpyrimidine derivatives as novel and potent PDE4 inhibitors.
PMID 24094436
Abstract
5-Carbamoyl-2-phenylpyrimidine derivative 2 has been identified as a phosphodiesterase 4 (PDE4) inhibitor with moderate PDE4B inhibitory activity (IC50=200 nM). Modification of the carboxylic acid moiety of 2 gave N-neopentylacetamide derivative 10f, which had high in vitro PDE4B inhibitory activity (IC50=8.3 nM) and in vivo efficacy against lipopolysaccharide (LPS)-induced pulmonary neutrophilia in mice (ID50=16 mg/kg, ip). Furthermore, based on the X-ray crystallography of 10f bound to the human PDE4B catalytic domain, we designed 7,8-dihydro-6H-pyrido[4,3-d]pyrimidin-5-one derivative 39 which has a fused bicyclic lactam scaffold. Compound 39 exhibited excellent inhibitory activity against LPS-induced tumor necrosis factor alpha (TNF-α) production in mouse splenocytes (IC50=0.21 nM) and in vivo anti-inflammatory activity against LPS-induced pulmonary neutrophilia in mice (41% inhibition at a dose of 1.0 mg/kg, i.t.).