EMAIL THIS PAGE TO A FRIEND
Nature chemical biology
Development of small-molecule inhibitors targeting adipose triglyceride lipase.
PMID 24096302
Abstract
Adipose triglyceride lipase (ATGL) is rate limiting in the mobilization of fatty acids from cellular triglyceride stores. This central role in lipolysis marks ATGL as an interesting pharmacological target as deregulated fatty acid metabolism is closely linked to dyslipidemic and metabolic disorders. Here we report on the development and characterization of a small-molecule inhibitor of ATGL. Atglistatin is selective for ATGL and reduces fatty acid mobilization in vitro and in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1075
Atglistatin, ≥98% (HPLC)
C17H21N3O
E0287 Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
E0162 Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
C4749 Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
E0412 Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
46051 Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
79302 Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥8.0 U/mg
96667 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
51963 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥0.8 U/mg
46069 Esterase from horse liver, lyophilized, powder, brown, 0.5-1.0 U/mg
46058 Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
E2884 Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
E3019 Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
E0887 Esterase from rabbit liver, lyophilized powder, ≥75 units/mg protein
79208 Esterase from Rhizopus oryzae, powder, light beige, ≥20 U/g
46071 Esterase from Saccharomyces cerevisiae, lyophilized, powder, beige, ~2 U/g
43388 Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
93855 Esterase Isoenzyme 2 porcine liver, recombinant, recombinant, expressed in E. coli, ≥80 U/g
93712 Esterase Isoenzyme 3 porcine liver, recombinant, recombinant, expressed in E. coli, ≥0.8 U/mg
68998 Esterase Isoenzyme 4 porcine liver, recombinant, recombinant, expressed in E. coli, ≥0.7 U/mg
53564 Esterase Isoenzyme 5 porcine liver, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
80712 Esterase Methylobacterium populi, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
92385 Esterase Nocardia farcinica, recombinant, recombinant, expressed in E. coli, ≥0.4 U/mg
04492 Esterase Paenibacillus barcinonensis, recombinant, recombinant, expressed in E. coli, ≥10 U/mg
04533 Esterase Pelobacter propionicus, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
75742 Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
80711 Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥0.3 U/mg
16698 Lipase, Candida antarctica, CLEA, ≥1.5 U/mg
62350 Lipozyme®, immobilized from Mucor miehei, granular, brown, >30 U/g