Description
Molecular Formula
Add to Cart
07-329
Anti-acetyl-Histone H4 (Lys16) Antibody, Anti-acetyl-Histone H4 (Lys16) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H4 (Lys16) also known as H4K16Ac, Histone H4 (acetyl K16 & has been published and validated in ChIP, WB, Mplex, PIA, DB, ChIP-seq.
07-327
Anti-acetyl-Histone H4 (Lys5) Antibody, Use Anti-acetyl-Histone H4 (Lys5) Antibody (Rabbit Polyclonal Antibody) validated in WB, DB, ICC, ChIP-seq, ChIP to detect acetyl-Histone H4 (Lys5) also known as H4K5Ac, Histone H4 (acetyl K5).
07-1584
Anti-dimethyl Histone H4 (Lys20) Antibody, Anti-dimethyl Histone H4 (Lys20) Antibody is a Rabbit Polyclonal Antibody for detection of dimethyl Histone H4 (Lys20) also known as H4K20me2, Histone H4 (di methyl K20) & has been validated in WB.
07-449
Anti-trimethyl-Histone H3 (Lys27) Antibody, Anti-trimethyl-Histone H3 (Lys27), also known as Anti-H3K27me3, is a highly published Rabbit Polyclonal Antibody. This protein A purified antibody is dot blot tested for trimethylated lysine 27 specificity and validated in WB, ICC, IP.
07-442
Anti-trimethyl-Histone H3 (Lys9) Antibody, Use Anti-trimethyl-Histone H3 (Lys9) Antibody (rabbit polyclonal antibody) validated in DB, ICC, Mplex, PIA, WB, ChIP-seq to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).
07-463
Anti-trimethyl-Histone H4 (Lys20) Antibody, Anti-trimethyl-Histone H4 (Lys20) Antibody is a rabbit polyclonal antibody for detection of trimethyl-Histone H4 (Lys20) also known as H4K20me3, Histone H4 (tri methyl K20) & has been validated in IP, WB, ChIP, DB.