EMAIL THIS PAGE TO A FRIEND
Annals of Indian Academy of Neurology
A Turkish family with Sjögren-Larsson syndrome caused by a novel ALDH3A2 mutation.
PMID 24101836
Abstract
Sjögren-Larsson syndrome (SLS) is an inherited neurocutaneous disorder caused by mutations in the aldehyde dehydrogenase family 3 member A2 (ALDH3A2) gene that encodes fatty aldehyde dehydrogenase. Affected patients display ichthyosis, mental retardation, and spastic diplegia. More than 70 mutations in ALDH3A2 have been discovered in SLS patients. We diagnosed two brothers age of 12 and 20 years with characteristic features of this rare syndrome. Magnetic resonance imaging showed demyelinating disease in both of them. We described a novel homozygous, c. 835 T > A (p.Y279N) mutation in exon 6 in two patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV44274
Anti-ALDH3A2 antibody produced in rabbit, affinity isolated antibody