EMAIL THIS PAGE TO A FRIEND
Retina (Philadelphia, Pa.)
Antibacterial properties of 2% lidocaine and reduced rate of endophthalmitis after intravitreal injection.
PMID 24105249
Abstract
To determine whether the application of subconjunctival 2% lidocaine/0.1% methylparaben for anesthesia may reduce rates of endophthalmitis after intravitreal (IVT) injection. We performed in vitro experiments to determine the antibacterial properties of 2% lidocaine/0.1% methylparaben (lidocaine) against causative organisms of endophthalmitis. Isolates of Staphylococcus aureus, Staphylococcus epidermidis, and Streptococcus viridans from patients with endophthalmitis were incubated with or without lidocaine. Aliquots (100 µL) were plated on Mueller-Hinton (S. aureus and S. epidermidis) or blood agar plates (S. viridans) at 0, 10, 30, 120, and 240 minutes, and colonies were counted after 24 hours. A retrospective review of 15,042 IVT injections was performed from January 2004 to February 2011 to determine the rate of endophthalmitis with or without application of subconjunctival lidocaine for anesthesia. Lidocaine demonstrated rapid bactericidal effects against all 3 organisms. After 10 minutes of exposure, there was approximately a 90% (P < 0.01), 95% (P < 0.001), and 92% (P < 0.001) reduction in colony forming units when compared with time 0 for S. aureus, S. epidermidis, and S. viridans, respectively. Complete elimination of colony forming units occurred at subsequent time points for each organism in contrast to logarithmic increase for control plates. There were a total of 0 cases of endophthalmitis of 6,853 IVT injections performed with subconjunctival lidocaine and 8 cases of endophthalmitis of 8,189 (0.1%) IVT injections performed with other methods of anesthesia (P = 0.03). Application of subconjunctival 2% lidocaine/0.1% methylparaben for anesthesia may reduce the incidence of endophthalmitis after IVT injection.