EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
SMG-1 suppresses CDK2 and tumor growth by regulating both the p53 and Cdc25A signaling pathways.
PMID 24107632
Abstract
The DNA damage response is coordinated by phosphatidylinositol 3-kinase-related kinases, ATM, ATR, and DNA-PK. SMG-1 is the least studied stress-responsive member of this family. Here, we show that SMG-1 regulates the G 1/S checkpoint through both a p53-dependent, and a p53-independent pathway. We identify Cdc25A as a new SMG-1 substrate, and show that cells depleted of SMG-1 exhibit prolonged Cdc25A stability, failing to inactivate CDK2 in response to radiation. Given an increased tumor growth following depletion of SMG-1, our data demonstrate a novel role for SMG-1 in regulating Cdc25A and suppressing oncogenic CDK2 driven proliferation, confirming SMG-1 as a tumor suppressor.