EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Effects of D,L-4-fluoroglutamic acid on glutamylation of methotrexate by hepatic cells in vitro.
PMID 2411272
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89377
4-Fluoro-DL-glutamic acid, ≥93%
C5H8FNO4