EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
Link up and fold up--templating the formation of spectrin tetramers.
PMID 24112938
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3644 Spectrin from human erythrocytes, buffered aqueous glycerol solution