EMAIL THIS PAGE TO A FRIEND
European Journal of Endocrinology
Longitudinal trends in thyroid function in relation to iodine intake: ongoing changes of thyroid function despite adequate current iodine status.
PMID 24114433