EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Nickel-catalyzed sonogashira reactions of non-activated secondary alkyl bromides and iodides.
PMID 24115611
Abstract
A nicked reaction: The title reaction of terminal alkynes with non-activated secondary alkyl iodides and bromides was accomplished for the first time. This reaction provides a new and practical approach for the synthesis of substituted alkynes (see scheme; cod=cyclo-1,5-octadiene).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
238244
Iodocyclohexane, 98%
C6H11I