EMAIL THIS PAGE TO A FRIEND
Dong wu xue yan jiu = Zoological research
[Perinatal clomiphene citrate treatment changes sexual orientations of male mice].
PMID 24115661
Abstract
Perinatal period and adolescence are critical for brain development, which is the biological basis of an individual's sexual orientation and sexual behavior. In this study, animals were divided into two groups and their sexual orientations were observed: one group experienced drug treatments during the perinatal period, and the other group was castrated at puberty. The results showed that estradiol treatment had no effect on mature male offspring's sexual orientations, but 9 days and 14 days of clomiphene citrate treatment significantly increased the chance of homosexuality and effeminized behavior. In addition, the sexual orientation of mature normal male offspring, which were castrated when they were 21 days old,was not significant different from the control animals. These findings suggest that the inhibition of perinatal estrogen activities could suppress individual male-typical responses, enhance female-typical responses and induce homosexual orientations. Moreover, the masculinizing effects of estrogen were more obvious during perinatal period than adolescence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0719 Enclomiphene hydrochloride, ≥96% (HPLC)
C26H28ClNO · HCl