EMAIL THIS PAGE TO A FRIEND
Dong wu xue yan jiu = Zoological research
The first mitochondrial genome for the butterfly family Riodinidae (Abisara fylloides) and its systematic implications.
PMID 24115668
Abstract
The Riodinidae is one of the lepidopteran butterfly families. This study describes the complete mitochondrial genome of the butterfly species Abisara fylloides, the first mitochondrial genome of the Riodinidae family. The results show that the entire mitochondrial genome of A. fylloides is 15 301 bp in length, and contains 13 protein-coding genes, 2 ribosomal RNA genes, 22 transfer RNA genes and a 423 bp A+T-rich region. The gene content, orientation and order are identical to the majority of other lepidopteran insects. Phylogenetic reconstruction was conducted using the concatenated 13 protein-coding gene (PCG) sequences of 19 available butterfly species covering all the five butterfly families (Papilionidae, Nymphalidae, Peridae, Lycaenidae and Riodinidae). Both maximum likelihood and Bayesian inference analyses highly supported the monophyly of Lycaenidae+Riodinidae, which was standing as the sister of Nymphalidae. In addition, we propose that the riodinids be categorized into the family Lycaenidae as a subfamilial taxon. The Riodinidae is one of the lepidopteran butterfly families. This study describes the complete mitochondrial genome of the butterfly species