EMAIL THIS PAGE TO A FRIEND
International Journal of Cancer. Journal International Du Cancer
Patterns of expression of keratin 19 as detected with monoclonal antibodies in human breast tissues and tumours.
PMID 2411673