EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Enrichment of phosphatidylinositols with specific acyl chains.
PMID 24120446
Abstract
There are six major species of phospholipids in eukaryotes, each of which plays unique structural and functional roles. One species, phosphatidylinositol (PI) only contributes about 2-10% of the total phospholipid pool. However, they are critical factors in the regulation of several fundamental processes such as in membrane dynamics and signal transduction pathways. Although numerous acyl species exist, PI species are enriched with one specific acyl chain composition at both sn-1 and sn-2 positions. Recent work has identified several enzymes that act on lipids to lead to the formation or interconversion of PI species that exhibit acyl chain specificity. These enzymes contribute to this lipid's enrichment with specific acyl chains. The nature of the acyl chains on signaling lipids has been shown to contribute to their specificity. Here we review some of the critical functions of PI and the multiple pathways in which PI can be produced and metabolized. We also discuss a common motif that may confer arachidonoyl specificity to several of the enzymes involved. This article is part of a Special Issue entitled: Membrane Structure and Function: Relevance in the Cell's Physiology, Pathology and Therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8798
Arachidonic acid sodium salt, from porcine liver, ≥99% (capillary GC), waxy solid
C20H31O2Na