EMAIL THIS PAGE TO A FRIEND
Neurobiology of Disease
Region-specific deficits in dopamine, but not norepinephrine, signaling in a novel A30P ?-synuclein BAC transgenic mouse.
PMID 24121116
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB152
Anti-Tyrosine Hydroxylase Antibody