EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Lichen planopilaris treated by the peroxisome proliferator activated receptor-γ agonist pioglitazone: lack of lasting improvement or cure in the majority of patients.
PMID 24124823