EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Risk of adverse effects, misdiagnosis, and suboptimal patient care with the use of over-the-counter triamcinolone. Con.
PMID 24125134