EMAIL THIS PAGE TO A FRIEND
Journal of biophotonics
Low-level laser therapy (810 nm) protects primary cortical neurons against excitotoxicity in vitro.
PMID 24127337
Abstract
Excitotoxicity describes a pathogenic process whereby death of neurons releases large amounts of the excitatory neurotransmitter glutamate, which then proceeds to activate a set of glutamatergic receptors on neighboring neurons (glutamate, N-methyl-D-aspartate (NMDA), and kainate), opening ion channels leading to an influx of calcium ions producing mitochondrial dysfunction and cell death. Excitotoxicity contributes to brain damage after stroke, traumatic brain injury, and neurodegenerative diseases, and is also involved in spinal cord injury. We tested whether low level laser (light) therapy (LLLT) at 810 nm could protect primary murine cultured cortical neurons against excitotoxicity in vitro produced by addition of glutamate, NMDA or kainate. Although the prevention of cell death was modest but significant, LLLT (3 J/cm(2) delivered at 25 mW/cm(2) over 2 min) gave highly significant benefits in increasing ATP, raising mitochondrial membrane potential, reducing intracellular calcium concentrations, reducing oxidative stress and reducing nitric oxide. The action of LLLT in abrogating excitotoxicity may play a role in explaining its beneficial effects in diverse central nervous system pathologies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1821
DAF-FM, ≥98% (HPLC)
C21H14F2N2O5
G0355000
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
C5H9NO4