EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Palladium-catalyzed synthesis of benzofurans and coumarins from phenols and olefins.
PMID 24127393
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
379875
Palladium(II) acetate, 99.98% trace metals basis
C4H6O4Pd