EMAIL THIS PAGE TO A FRIEND
Stroke
Response to letter regarding article, "Asymmetric dimethylarginine in response to recombinant tissue-type plasminogen activator and erythropoietin in acute stroke".
PMID 24130135