EMAIL THIS PAGE TO A FRIEND
Human & Experimental Toxicology
Octanal-induced inflammatory responses in cells relevant for lung toxicity: expression and release of cytokines in A549 human alveolar cells.
PMID 24130214