EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Study on the chemical constituent from the dichloromethane extract of the pine needle of Cedrus deodara (II)].
PMID 24134002
Abstract
To study the chemical constituents of the dichloromethane extract from pine needles of Cedrus deodara. The chemical constituents were isolated and purified from the dichloromethane extract of pine needles by chromatography on silica gel and Sephadex LH-20. Their structures were elucidated by analysis of spectral data and chemical properties. Seven compounds were isolated and their chemical structures were identified as ferulic acid (1), osthole (2), beta-phenylacrylic acid (3), paeonol (4), beta-sitosterol (5), magnolol (6) and honokiol (7). Compounds 1 - 4, 6 and 7 are isolated from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90034
4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical standard
C10H10O4