EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
UHPLC-MS/MS analysis of arachidonic acid and 10 of its major cytochrome P450 metabolites as free acids in rat livers: effects of hepatic ischemia.
PMID 24134894