EMAIL THIS PAGE TO A FRIEND
Molecular Carcinogenesis
Gestational exposure to the AhR agonist 2,3,7,8-tetrachlorodibenzo-p-dioxin induces BRCA-1 promoter hypermethylation and reduces BRCA-1 expression in mammary tissue of rat offspring: preventive effects of resveratrol.
PMID 24136580