EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Thiophosphoramide-based cooperative catalysts for Brønsted acid promoted ionic Diels-Alder reactions.
PMID 24136865
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
803863
Schreiner′s Thiourea Catalyst, 95%
C17H8F12N2S