EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Conjugates of cytochrome
PMID 24137266
Abstract
Polycationic cell-penetrating peptides (CPPs) deliver macromolecules into cells without losing the functional properties of the cargoed macromolecule. The aim of this study was to determine whether exogenous cytochrome
Related Materials