EMAIL THIS PAGE TO A FRIEND
Ultrasonics
Surface and quasi-longitudinal acoustic waves in KTiOAsO₄ single crystals.
PMID 24139300
Abstract
Surface and quasi-longitudinal acoustic wave properties have been investigated in potassium titanyl arsenate (KTiOAsO₄, KTA) single crystals for the first time. Surface acoustic wave (SAW) velocity, electromechanical coupling coefficient and power flow angle characteristics have been obtained in rotated Y-cut of KTA crystals. High SAW electromechanical coupling coefficient (0.4%) is found in Z-cut of KTA crystals. For high-frequency devices it is promising the resonators on quasi-longitudinal acoustic wave in X-cut of KTA crystals with sharp response in interdigital transducer conductance at resonance frequency.