EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Magnetic-particle-based, ultrasensitive chemiluminescence enzyme immunoassay for free prostate-specific antigen.
PMID 24139579
Abstract
We report a magnetic-particle (MMP)-based chemiluminescence enzyme immunoassay (CLEIA) for free prostate-specific antigen (f-PSA) in human serum. In this method, the f-PSA is sandwiched between the anti-PSA antibody coated MMPs and alkaline phosphatase (ALP)-labeled anti-f-PSA antibody. The signal produced by the emitted photons from the chemiluminescent substrate (4-methoxy-4-(3-phosphatephenyl)-spiro-(1,2-dioxetane-3,2'-adamantane)) is directly proportional to the amount of f-PSA in a sample. The present MMP-based assay can detect f-PSA in the range of 0.1-30 ng mL(-1) with the detection limit of 0.1 ng mL(-1). The linear detection range could match the concentration range within the "diagnostic gray zone" of serum f-PSA levels (4-10 ng mL(-1)). The detection limit was sufficient for measuring clinically relevant f-PSA levels (>4 ng mL(-1)). Furthermore, the method was highly selective; it was unaffected by cross-reaction with human glandular kallikrein-2, a kallikrein-like serine protease that is 80% similar to f-PSA. The proposed method was finally applied to determine f-PSA in 40 samples of human sera. Results obtained using the method showed high correlation with those obtained using a commercially available microplate CLEIA kit (correlation coefficient, 0.9821). This strategy shows great potential application in the fabrication of diagnostic kits for determining f-PSA in serum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K3627 Kallikrein from porcine pancreas, ≥40 units/mg protein